Learn More
Aviva Systems Biology Corp SLC7A1 Antibody - N-terminal region (ARP43838_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43838_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC7A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC7A1. Alternate names: ATRC1, CAT-1, ERR, HCAT1, REC1LApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-014-53379