Learn More
Aviva Systems Biology Corp SLC7A11 Antibody - middle region (ARP43947_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43947_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC7A11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC7A11. Alternate names: CCBR1, xCTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58672