Learn More
Aviva Systems Biology Corp SLC7A14 Antibody - N-terminal region (ARP44014_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44014_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC7A14. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC7A14. Alternate names: KIAA1613ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53392