Learn More
Aviva Systems Biology Corp SLC7A2 Antibody - middle region (ARP43766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC7A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC7A2. Alternate names: ATRC2, CAT-2, HCAT2, CAT2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58624