Learn More
Aviva Systems Biology Corp Slc7a3 Antibody - middle region (ARP43806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc7a3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Atrc3, CAT3, SLC7A1, SLC7A2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-099