Learn More
Aviva Systems Biology Corp SLC7A5 Antibody - N-terminal region (ARP63651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC7A5. It was validated on Western Blot. . Alternate names: 4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1ApplicationWB. Species reactivity: Human
Catalog No. 50-288-043