Learn More
Aviva Systems Biology Corp SLC9A3 Antibody - middle region (ARP43870_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43870_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC9A3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SLC9A3. Alternate names: MGC126718, MGC126720, NHE3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-104