Learn More
Aviva Systems Biology Corp Slc9a7 Antibody - C-terminal region (ARP58392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc9a7. It was validated on Western Blot. . Alternate names: A530087D17Rik, NHE7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-471