Learn More
Aviva Systems Biology Corp SLC9A7 Antibody - N-terminal region (ARP44066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC9A7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC9A7. Alternate names: NHE7, SLC9A6ApplicationWB. Species reactivity: Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-58706