Learn More
Med Vet International Closed Wound Silicone Drainage Kit, 100mL, J0790
Catalog No. 5000495298
Encompass_Preferred
This vacuum drain‘s most common use is for post-op drainage Jackson Pratt. The kit is designed to create suction or negative pressure for draining wounds. A fenestrated silicone drain is placed into position with the aid of a stainless steel trocar. This drain is attached to a silicone bulb with a one-way valve. When the bulb is released a vacuum is created that pulls exudates out through the drain. Wound exudates can be stored, visualized, or cultured. The drain has a radio-opaque line for the entire length.The kit consists of: a 130mm stainless steel trocar, silicone fenestrated drain and tubing, a silicone bulb and a clamp for attachment to bandage or patient.
Catalog No. 50-004-95298 Supplier Med Vet International Supplier No. JOR790
This vacuum drain‘s most common use is for post-op drainage Jackson Pratt. The kit is designed to create suction or negative pressure for draining wounds. A fenestrated silicone drain is placed into position with the aid of a stainless steel trocar. This drain is attached to a silicone bulb with a one-way valve. When the bulb is released a vacuum is created that pulls exudates out through the drain. Wound exudates can be stored, visualized, or cultured. The drain has a radio-opaque line for the entire length.The kit consists of: a 130mm stainless steel trocar, silicone fenestrated drain and tubing, a silicone bulb and a clamp for attachment to bandage or patient.