Learn More
Aviva Systems Biology Corp SLCO1A2 Antibody - middle region (ARP43899_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43899_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO1A2. It was validated on Western Blot. SLCO1A2,solute carrier organic anion transporter family, member 1A2,OATP; OATP-A; OATP1A2; SLC21A3; SLCO1A2,solute carrier organic anion transporter family member 1A2,solute carrier organic anion transporter family member 1A2,OATP-1,OTTHUMP00000209605,solute carrier family 21 member 3,organic anion transporting polypeptide A,organic anion-transporting polypeptide 1,sodium-independent organic anion transporter,solute carrier family 21 (organic anion transporter), member 3The immunogen is a synthetic peptide directed towards the middle region of human SLCO1A2. Alternate names: OATP, OATP-A, OATP1A2, SLC21A3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-109