Learn More
Aviva Systems Biology Corp SLCO1B1 Antibody - middle region (ARP43914_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43914_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO1B1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SLCO1B1. Alternate names: LST-1, MGC133282, OATP-C, OATP1B1, OATP2, SLC21A6, LST1, HBLRR, OATPCApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-285-110