Learn More
Aviva Systems Biology Corp SLCO1C1 Antibody - N-terminal region (ARP43970_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO1C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLCO1C1. Alternate names: OATP-F, OATP1, OATP1C1, SLC21A14, OATPF, OATP14, OATPRP5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58678