Learn More
Biomatik Corporation Slco2A1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11585
Encompass_Preferred
Slco2A1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLCO2A1. The Slco2A1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Organic Anion Transporter Family Member 2A1 Protein (416-518Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLCO2A1. Accession Number: Q92959. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5204