Learn More
Aviva Systems Biology Corp SLCO2B1 Antibody - N-terminal region (ARP43926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLCO2B1. Alternate names: DKFZp686E0517, KIAA0880, OATP-B, OATP2B1, OATPB, SLC21A9ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53383