Learn More
Aviva Systems Biology Corp SLCO3A1 Antibody - middle region (ARP43934_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO3A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLCO3A1. Alternate names: FLJ40478, OATP-D, OATP3A1, SLC21A11, OATPDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53387