Learn More
Aviva Systems Biology Corp SLCO5A1 Antibody - middle region (ARP44052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLCO5A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SLCO5A1. Alternate names: FLJ39560, OATP-J, OATP5A1, OATPRP4, SLC21A15, OATP-RP4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-121