Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Masterflex Spooled Pump Tubing, Platinum-Cured Silicone, L/S 24, 300 ft

Supplier:  Accuflow Systems 9640324

Encompass_Preferred

Masterflex L/S 24 spooled platinum-cured silicone pump tubing provides continuous lengths for precise peristaltic and fluid transfer in pharmaceutical, biotech, food, and laboratory applications.

  • Platinum-cured silicone material.
  • L/S 24 size, hose barb 1/4 in (6.4 mm).
  • 300 ft (91.4 m) spooled continuous length.
  • Temperature range -50 to 230 °C (-58 to 446 °F).
  • Compliant with FDA 21 CFR 177.2600, USP, European Pharmacopoeia, 3A, cGMP, RoHS, ADCF, and REACH (non-DEHP).
  • Spooled packaging reduces scrap and eases storage and handling.

Catalog No. NC1563743


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.