Learn More
Research Products International Corp Smart Slicer Blade, 25 x 50 x 0.22mm, 100/pk
SDP
Supplier:  Research Products International Corp 246811
Encompass
SmartSlicer provides a unique alternative to single edge razor blades. Made from ultra thin (0.22 mm) food grade PET plastic that performs like a conventional blade, however is much safer to use. Cleanly slice right on the light box without scratching the surface. Ideal for a variety of laboratory applications including slicing gels, soft tissue, plant matter and more. Adjust the size required by simply cutting with scissors. Made in the USA.
Catalog No. 50-222-9625