Learn More
Cole-Parmer Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 6.5; 15/Pk
Catalog No. NC1595213 Shop All Cole-Parmer Products
Encompass_Preferred
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Catalog No. NC1595213 Supplier Cole-Parmer Supplier No. 6299120
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.