Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Polysine™Microscope Adhesion Slides

Catalog No. 1254578 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W):
75 x 25 mm
Type:
Polysine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Type
12-545-78 75 x 25 mm Polysine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-545-78 Supplier Epredia™ Supplier No. P4981001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate the need for extra adhesive or coating techniques with Epredia™ Polysine Microscope Adhesion Slides.

  • Not affected by chemicals or predigestion
  • Ideal for immunocytochemical hybridization assays on cell preparation or tissue sections
  • Binds specimen to the slide throughout processing
  • High purity, corrosion-resistant white glass adhesion slide
  • Manufactured from pure, water-white glass
  • Pre-cleaned for immediate use
  • Packed 72 slides per box, 20 boxes per case

Specifications

Length (Metric) 75 mm
Type Polysine
Material Polysine
Edges Ground
Width (Metric) 25 mm
Precleaned Yes
Frosted Yes
Thickness (Metric) 1 mm
Color White
Corner Style 90°
Dimensions (L x W) 75 x 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.