Learn More
Medicus Health Slide Staining Plastic Dipper With Handle
SDP
Catalog No. NC1862867
Encompass
∙ Slide Staining Dipper is great for histological, cytological and hematological staining procedures with total immersion for slides∙Great for Hematology Departments who need to do Wright's Stains∙Plastic staining dipper holds maximum of 25 slides∙Slide Staining Dipper is sold by the each∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H∙Staining Dipper is designed for 75mm x 25mm or 3" x 1" slides
Catalog No. NC1862867 Supplier Medicus Health Supplier No. 2935M6
∙ Slide Staining Dipper is great for histological, cytological and hematological staining procedures with total immersion for slides∙Great for Hematology Departments who need to do Wright's Stains∙Plastic staining dipper holds maximum of 25 slides∙Slide Staining Dipper is sold by the each∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H∙Staining Dipper is designed for 75mm x 25mm or 3" x 1" slides