Learn More
Medicus Health Slide Staining Station, 12 Place, 12 Dishes With Lids & Slide Dipper
SDP
Catalog No. NC1862866
Encompass
∙ Slide Staining 12 Place Station, Twelve Dishes with Lids and Slide Dipper ∙Slide Staining Station has (12) dishes with lids, (1) plastic slide dipper and (1) twelve bank stainless steel dish holding rack∙Twelve Place Staining Station great for busy Hematology Departments who need to do a quick Wright's Stain∙Compact, lightweight rack makes it easy to move and reposition∙Multiple units can be placed in a row to build any specialty staining procedure∙Dish capacity is 300 mL and dish is resistant to acids and solvents∙Staining Station is designed for 75mm x 25mm or 3" x 1" slides∙Plastic staining dipper holds maximum of 24 slides∙Staining Set Overall Dimensions: 28"W x 5"D x 5"H∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H
Catalog No. NC1862866 Supplier Medicus Health Supplier No. 2935M7
∙ Slide Staining 12 Place Station, Twelve Dishes with Lids and Slide Dipper ∙Slide Staining Station has (12) dishes with lids, (1) plastic slide dipper and (1) twelve bank stainless steel dish holding rack∙Twelve Place Staining Station great for busy Hematology Departments who need to do a quick Wright's Stain∙Compact, lightweight rack makes it easy to move and reposition∙Multiple units can be placed in a row to build any specialty staining procedure∙Dish capacity is 300 mL and dish is resistant to acids and solvents∙Staining Station is designed for 75mm x 25mm or 3" x 1" slides∙Plastic staining dipper holds maximum of 24 slides∙Staining Set Overall Dimensions: 28"W x 5"D x 5"H∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H