Learn More
Newcomer Supply Slide Staining Trays - Disposable. Holds up to 8 slides and comes with 4 bases and 1 lid.
SDP
Catalog No. NC0754694
Encompass
A durable constructed plastic slide staining tray that holds 8 slides comfortably. The deep well holds liquid waste up to 38ml. The tray provides easy transport from work area to sink. Two convenient pour spouts make it easy to dispose of unwanted liquid waste. Dark lid & base protects slides when used for light sensitive applications. Trays are suitable for temps of -10°C to +80°C.
Catalog No. NC0754694 Supplier Newcomer Supply Supplier No. 6846
A durable constructed plastic slide staining tray that holds 8 slides comfortably. The deep well holds liquid waste up to 38ml. The tray provides easy transport from work area to sink. Two convenient pour spouts make it easy to dispose of unwanted liquid waste. Dark lid & base protects slides when used for light sensitive applications. Trays are suitable for temps of -10°C to +80°C.