Learn More
Medicus Health Slide Staining Station Dish With Lid
SDP
Catalog No. NC1969285
Encompass
∙ Slide Staining Station Dish with Lid is great for histological, cytological and hematological staining procedures with total immersion for slides∙Great for Hematology Departments who need to do Wright's Stains∙Dish and Lid are sold together∙Multiple units can be placed in a row to build any specialty staining procedure∙Stain Dish with lids to help reduce reagent evaporation or contamination ∙Dish capacity is 300 mL and dish is resistant to acids and solvents∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides∙Dish measures: 4"L x 2"W x 4"H
This item is not returnable. View return policy
Catalog No. NC1969285 Supplier Medicus Health Supplier No. 2935M5
∙ Slide Staining Station Dish with Lid is great for histological, cytological and hematological staining procedures with total immersion for slides∙Great for Hematology Departments who need to do Wright's Stains∙Dish and Lid are sold together∙Multiple units can be placed in a row to build any specialty staining procedure∙Stain Dish with lids to help reduce reagent evaporation or contamination ∙Dish capacity is 300 mL and dish is resistant to acids and solvents∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides∙Dish measures: 4"L x 2"W x 4"H