Learn More
Cole-Parmer Essentials Slideable Sterile Petri Dishes, 100 x 15 mm; 500/CS
Catalog No. NC2097006 Shop All Cole-Parmer Products
Encompass_Preferred
A partial edge ring on base allows a single dish to be slid from a large stack and enhances stability when stacked. Consistent flatness provides for even media distribution. Dishes are made of virgin polystyrene and are optically clear for improved imaging and observation. All dishes are supplied sterile (gamma irradiated) and come packaged 20 dishes/bag.
This item is not returnable. View return policy
Catalog No. NC2097006 Supplier Cole-Parmer Supplier No. 0193231
A partial edge ring on base allows a single dish to be slid from a large stack and enhances stability when stacked. Consistent flatness provides for even media distribution. Dishes are made of virgin polystyrene and are optically clear for improved imaging and observation. All dishes are supplied sterile (gamma irradiated) and come packaged 20 dishes/bag.