Learn More
Waters Corp Sleeve, 2 mL Vial, 4/pk
Supplier:  Waters Corp 700005338
Encompass_Preferred
This sleeve or vial holder accommodates 2mL vials within the standard 4mL auxiliary vial positions on the sample manager shuttle tray or rotary platter. This can be used in all ACQUITY Sample Managers including the ACQUITY Arc System
Catalog No. 50-146-2035