Learn More
Ace Glass, Inc. SLEEVE-34/45 PTFE PK/12 SLEEVE-34/45 PTFE PK/12
SDP
Catalog No. NC0040521
Encompass
PTFE sleeves are inserted between standard taper ground glass joints. Precludes contamination from lubricant or from leaks. They have a wall thickness of about 0.05mm, are inert, usable to 280 degrees C and will withstand low to medium vacuum.
Catalog No. NC0040521 Supplier Ace Glass, Inc. Supplier No. 7643112
PTFE sleeves are inserted between standard taper ground glass joints. Precludes contamination from lubricant or from leaks. They have a wall thickness of about 0.05mm, are inert, usable to 280 degrees C and will withstand low to medium vacuum.