Learn More
Sheldon Manufacturing Inc SLEEVE ASSEMBLY 1 PR
SDP
Catalog No. NC0729748
Encompass_Preferred
One pair of sleeves for Bactron or Bactrox chambers with 10" ergonomic armports. Does not contain cuffs, cuff rings, or o-rings. See 9990738S,M or L for standard assemblies, or 9990812S, M, or L for latex-free assemblies.
This item is not returnable. View return policy
Catalog No. NC0729748 Supplier Sheldon Manufacturing Inc Supplier No. 3600521
One pair of sleeves for Bactron or Bactrox chambers with 10" ergonomic armports. Does not contain cuffs, cuff rings, or o-rings. See 9990738S,M or L for standard assemblies, or 9990812S, M, or L for latex-free assemblies.