Learn More
Sheldon Manufacturing Inc SLEEVE ASSEMBLY ERGO SIZE 8
SDP
Catalog No. NC0945945
Encompass_Preferred
Sleeve assembly for Bactron anaerobic chambers with ergonomic 10" diamater armports. Medium, size 8. Includes sleeves, cuffs, cuff rings, and o-rings pre-assembled.
This item is not returnable. View return policy
Catalog No. NC0945945 Supplier Sheldon Manufacturing Inc Supplier No. 9990738M
Sleeve assembly for Bactron anaerobic chambers with ergonomic 10" diamater armports. Medium, size 8. Includes sleeves, cuffs, cuff rings, and o-rings pre-assembled.