Learn More
Sheldon Manufacturing Inc SLEEVE ASSEMBLY LG
SDP
Catalog No. NC0900690
Encompass_Preferred
Sleeve assembly for legacy Bactron anaerobic chambers with round 8" diamater armports. Large, Size 9. Includes sleeves, cuffs, cuff rings, and o-rings pre-assembled.
This item is not returnable. View return policy
Catalog No. NC0900690 Supplier Sheldon Manufacturing Inc Supplier No. 9990514L
Sleeve assembly for legacy Bactron anaerobic chambers with round 8" diamater armports. Large, Size 9. Includes sleeves, cuffs, cuff rings, and o-rings pre-assembled.