Learn More
Waters Corp ESI Conductive Sleeve
Catalog No. 50819194 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ESI Conductive Sleeve is made of foam material and it holds the ESI stainless steel capillary in place on the probe.
Catalog No. 50-819-194 Supplier Waters Corp Supplier No. 700000969
The ESI Conductive Sleeve is made of foam material and it holds the ESI stainless steel capillary in place on the probe.