Learn More
IDEX Health & Science Upchurch SLEEVE PEEK F/.030 OD TUBING
Catalog No. NC1919740
Encompass_Preferred
Idex Tubing Sleeves, Natural, Diameter Inner: 785 um, For Use With: 730 to 770 um O.D. Tubing, PEEK, Diameter Outer: 0.265 in., 1.57 cm
Catalog No. NC1919740 Supplier IDEX Health & Science Upchurch Supplier No. F232
Idex Tubing Sleeves, Natural, Diameter Inner: 785 um, For Use With: 730 to 770 um O.D. Tubing, PEEK, Diameter Outer: 0.265 in., 1.57 cm