Learn More
Wilmad Labglass SLEEVE PTFE ST 29/42
Catalog No. 50802131
Encompass
Joint Sleeve, PTFE. Joint Size: 29/42. Joint sleeves are fitted onto standard taper inner joints to prevent freezing of assembled joints. Sleeve nominal thickness is 0.003 inch.
Catalog No. 50-802-131 Supplier Wilmad Labglass Supplier No. LG1038T118
Joint Sleeve, PTFE. Joint Size: 29/42. Joint sleeves are fitted onto standard taper inner joints to prevent freezing of assembled joints. Sleeve nominal thickness is 0.003 inch.