Learn More
Supelco Inc SLEEVE STOPPERS 100/PK
Catalog No. NC9073710
Encompass
Sleeve stopper septa, bottom I.D. A- O.D. 9.3A mm A- 15.7A mm, white
Catalog No. NC9073710 Supplier Supelco Inc Supplier No. Z564729100EA
Sleeve stopper septa, bottom I.D. A- O.D. 9.3A mm A- 15.7A mm, white