Learn More
StatLab Sleeved Embedding Cassettes
SDP
Catalog No. 21060038
Change view
Click to view available options
Color:
Gray
Catalog No. Color
21-060-038 Gray
1 options
This item is not returnable. View return policy
Catalog No. 21-060-038 Supplier StatLab Supplier No. M4859SL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unique Front-hinged Design.
  • Unique front-hinged routine cassette design with a rear tab that allows for unobstructed access to the front labeling surface
  • Sleeved with lids separate
  • 45° labeling surface
  • 750/Cs.
Specifications
Color Gray
Product Type Embedding Cassettes