Learn More
Medchemexpress LLC HY-114872 10mg , SLF CAS:195513-96-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11487210MG
Encompass_Preferred
HY-114872 10mg SLF CAS: 195513-96-3 SLF is a synthetic ligand for FK506-binding protein (FKBP) with an affinity of 3.1 μM for FKBP51 and an IC50 of 2.6 μM for FKBP12. SLF can be used in the synthesis of PROTAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0570