Learn More
Medchemexpress LLC HY-107452 100mg Medchemexpress, SLF-amido-C2-COOH CAS:1092369-24-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107452100MG
Encompass_Preferred
Medchemexpress, HY-107452 100mg SLF-amido-C2-COOH CAS:1092369-24-8 SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) is a synthetic ligand for FKBP (SLF). SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9594