Learn More
Medchemexpress LLC SLF-amido-C2-COOH 25mg | 1092369-24-8 | 25MG
SDP
Catalog No. 5000241505
Encompass_Preferred
SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) is a synthetic ligand for FKBP (SLF) SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) can be used in the synthesis of PROTACs[1
Catalog No. 50-002-41505 Supplier Medchemexpress LLC Supplier No. HY10745225MG
Add to Cart
Add to Cart
SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) is a synthetic ligand for FKBP (SLF) SLF-amido-C2-COOH (PROTAC FKBP12-binding moiety 1) can be used in the synthesis of PROTACs[1