Learn More
Cayman Chemical SLF1081851hydrochlorIde 10mg
SDP
Catalog No. 502915037
Encompass_Preferred
An inhibitor of SPNS2; inhibits mouse Spns2 by 67% at 2 µM; inhibits S1P export from HeLa cells expressing mouse Spns2 (IC50 = 1.93 µM); decreases blood lymphocyte numbers and plasma S1P levels in mice at 20 mg/kg
Catalog No. 50-291-5037 Supplier Cayman Chemical Supplier No. 3715010 MG
An inhibitor of SPNS2; inhibits mouse Spns2 by 67% at 2 µM; inhibits S1P export from HeLa cells expressing mouse Spns2 (IC50 = 1.93 µM); decreases blood lymphocyte numbers and plasma S1P levels in mice at 20 mg/kg