Learn More
McKesson General Medical Sulfolyser Hematology Reagent, Enhances Cell Counting, Saves Processing Time, 2 x 1.5 L
Catalog No. NC2143721
Encompass_Preferred
The Sulfolyser Hematology Reagent is designed for precise hemoglobin measurement with Sysmex automated analyzers. This cyanide-free reagent enhances cell counting and supports differential analysis, ensuring rapid erythrocyte lysis and minimal errors. Its optimized formula contributes to efficient hematology testing, reducing processing time and improving accuracy, making it a reliable choice for clinical laboratories.
This item is not returnable. View return policy
Catalog No. NC2143721 Supplier McKesson General Medical Supplier No. 1192822
The Sulfolyser Hematology Reagent is designed for precise hemoglobin measurement with Sysmex automated analyzers. This cyanide-free reagent enhances cell counting and supports differential analysis, ensuring rapid erythrocyte lysis and minimal errors. Its optimized formula contributes to efficient hematology testing, reducing processing time and improving accuracy, making it a reliable choice for clinical laboratories.