Learn More
Enterprise Technology Solutions Slice, Inc Ceramic Blade, Rounded Tip, 4/5"x2-2/5"x1/10", 2/PK, White
SDP
Catalog No. 5000829127
Encompass_Preferred
Utility blade is made with 100 percent zirconium oxide, an advanced ceramic material, to provide a number of useful features that set it apart from typical steel blades. Zirconium oxide is nonsparking, nonconductive, nonmagnetic, chemically inert and never rusts. Slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Its natural edge retention utilizes a finger-friendly grind to resist piercing skin while maintaining its high-quality cutting performance. Plus, its safer edge eliminates the need for special sharps disposal boxes. Reversible blade with round tip is designed for use Slice 10490 and 10491 handles. It is also 100 percent recyclable.
Catalog No. 50-008-29127 Supplier Enterprise Technology Solutions Supplier No. SLI10524
Utility blade is made with 100 percent zirconium oxide, an advanced ceramic material, to provide a number of useful features that set it apart from typical steel blades. Zirconium oxide is nonsparking, nonconductive, nonmagnetic, chemically inert and never rusts. Slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Its natural edge retention utilizes a finger-friendly grind to resist piercing skin while maintaining its high-quality cutting performance. Plus, its safer edge eliminates the need for special sharps disposal boxes. Reversible blade with round tip is designed for use Slice 10490 and 10491 handles. It is also 100 percent recyclable.