Learn More
Enterprise Technology Solutions Slice, Inc Ceramic Blades,Pointed Tip,Dual,1/20"x1-3/10"x1/4",4/PK,WE
SDP
Catalog No. 5000790406
Encompass_Preferred
Box cutter blade is made with 100 percent zirconium oxide, an advanced ceramic material, to provide a number of useful features that set it apart from typical steel blades. Zirconium oxide is nonsparking, nonconductive and nonmagnetic. It is chemically inert and never rusts. Slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Its natural edge retention utilizes a finger-friendly grind to resist piercing skin while maintaining its high-quality cutting performance. Plus, its safer edge eliminates the need for special sharps disposal boxes. Pointed tip is perfect for applications that require piercing to initiate the cut. Reversible blade is designed for use with all Slice box cutters, pen cutters and the Mini cutter (none included).
Catalog No. 50-007-90406 Supplier Enterprise Technology Solutions Supplier No. SLI10408
Box cutter blade is made with 100 percent zirconium oxide, an advanced ceramic material, to provide a number of useful features that set it apart from typical steel blades. Zirconium oxide is nonsparking, nonconductive and nonmagnetic. It is chemically inert and never rusts. Slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Its natural edge retention utilizes a finger-friendly grind to resist piercing skin while maintaining its high-quality cutting performance. Plus, its safer edge eliminates the need for special sharps disposal boxes. Pointed tip is perfect for applications that require piercing to initiate the cut. Reversible blade is designed for use with all Slice box cutters, pen cutters and the Mini cutter (none included).