Learn More
Enterprise Technology Solutions Slice, Inc Utility Knife, Auto-Retract, 9/10"Wx6-1/10"Lx1-2/5"H, GYGN
SDP
Catalog No. 5000829131
Encompass_Preferred
Utility knife features a long blade design to efficiently cut through thick materials, including triple-wall corrugated. The advanced ceramic material is rustproof, nonconductive, antimagnetic and won't spark. The slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Auto-retractable blade stays out of harm's way when you're not using the knife for safe storage. Ambidextrous design provides a comfortable grip for both left-handed and right-handed users. Easy, no-tool blade change allows you to simply replace dull blades without requiring any extra tools. Lanyard hole allows you to keep this unique tool in a convenient location to ensure it's always close at hand. Utility knife includes one double-sided, rounded-tip safety blade.
Catalog No. 50-008-29131 Supplier Enterprise Technology Solutions Supplier No. SLI10554
Utility knife features a long blade design to efficiently cut through thick materials, including triple-wall corrugated. The advanced ceramic material is rustproof, nonconductive, antimagnetic and won't spark. The slow-to-dull blade starts at an effective sharpness and retains its safe cutting edge for up to 11 times longer than steel for consistently smooth cuts again and again. Auto-retractable blade stays out of harm's way when you're not using the knife for safe storage. Ambidextrous design provides a comfortable grip for both left-handed and right-handed users. Easy, no-tool blade change allows you to simply replace dull blades without requiring any extra tools. Lanyard hole allows you to keep this unique tool in a convenient location to ensure it's always close at hand. Utility knife includes one double-sided, rounded-tip safety blade.