Learn More
Hampton Research SLICE PH REAGENT #06 (A6)
SDP
Catalog No. 50562864
Encompass
HR2-070-06/Slice pH #06 (A6), 185 ml - 1.0 M Sodium citrate tribasic dihydrate pH 3.9. Please note items from this supplier are non-returnable.
Catalog No. 50-562-864 Supplier Hampton Research Supplier No. HR207006
HR2-070-06/Slice pH #06 (A6), 185 ml - 1.0 M Sodium citrate tribasic dihydrate pH 3.9. Please note items from this supplier are non-returnable.