Learn More
Hampton Research SLICE PH REAGENT #07 (A7)
SDP
Catalog No. 50562865
Encompass
HR2-070-07/Slice pH #07 (A7), 185 ml - 1.0 M Sodium citrate tribasic dihydrate pH 4.2. Please note items from this supplier are non-returnable.
Catalog No. 50-562-865 Supplier Hampton Research Supplier No. HR207007
HR2-070-07/Slice pH #07 (A7), 185 ml - 1.0 M Sodium citrate tribasic dihydrate pH 4.2. Please note items from this supplier are non-returnable.