Learn More
Hampton Research SLICE PH REAGENT #11 (A11)
SDP
Catalog No. 50562869
Encompass
HR2-070-11/Slice pH #11 (A11), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.3. Please note items from this supplier are non-returnable.
Catalog No. 50-562-869 Supplier Hampton Research Supplier No. HR207011
HR2-070-11/Slice pH #11 (A11), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.3. Please note items from this supplier are non-returnable.