Learn More
Hampton Research SLICE PH REAGENT #12 (A12)
SDP
Catalog No. 50562870
Encompass
HR2-070-12/Slice pH #12 (A12), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-562-870 Supplier Hampton Research Supplier No. HR207012
HR2-070-12/Slice pH #12 (A12), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.