Learn More
Hampton Research SLICE PH REAGENT #13 (B1)
SDP
Catalog No. 50562871
Encompass
HR2-070-13/Slice pH #13 (B1), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.9. Please note items from this supplier are non-returnable.
Catalog No. 50-562-871 Supplier Hampton Research Supplier No. HR207013
HR2-070-13/Slice pH #13 (B1), 185 ml - 1.0 M Sodium acetate trihydrate pH 4.9. Please note items from this supplier are non-returnable.