Learn More
Hampton Research SLICE PH REAGENT #16 (B4)
SDP
Catalog No. 50562874
Encompass
HR2-070-16/Slice pH #16 (B4), 185 ml - 1.0 M DL-Malic acid pH 5.3. Please note items from this supplier are non-returnable.
Catalog No. 50-562-874 Supplier Hampton Research Supplier No. HR207016
HR2-070-16/Slice pH #16 (B4), 185 ml - 1.0 M DL-Malic acid pH 5.3. Please note items from this supplier are non-returnable.